4-(Butylcarbamoyl)-3-fluorphenylboronsäure 98%
Lieferant: Apollo Scientific
APOSPC3622-1GEA
198
EUR
APOSPC3622-1G
4-(Butylcarbamoyl)-3-fluorphenylboronsäure 98%
4-(Butylcarbamoyl)-3-fluorphenylboronsäure
Formel:
C₁₁H₁₅BFNO₃ Schmelzpunkt: 130…133 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08689527 CAS-Nummer: 874289-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...