4-Fluor-2-(trifluormethoxy)benzylbromid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
APOSPC302591-1GEA
161
EUR
APOSPC302591-1G
4-Fluor-2-(trifluormethoxy)benzylbromid 98%
4-Fluor-2-(trifluormethoxy)benzylbromid
Formel:
C₈H₅BrF₄O Molecular Weight: 273,02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19687175 CAS-Nummer: 1323966-22-8 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...