3-(4-(Difluormethoxy)phenyl)propionsäure 97%
Lieferant: Apollo Scientific
APOSPC302513-1GEA
118
EUR
APOSPC302513-1G
3-(4-(Difluormethoxy)phenyl)propionsäure 97%
3-(4-(Difluormethoxy)phenyl)propionsäure
Formel:
C₁₀H₁₀F₂O₃ Molecular Weight: 216,18 g/mol Schmelzpunkt: 66…69 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09863355 CAS-Nummer: 1000932-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...