3',5'-Difluor-2'-hydroxyacetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC2826-1GEA
16.1
EUR
APOSPC2826-1G
3',5'-Difluor-2'-hydroxyacetophenon 98%
3',5'-Difluor-2'-hydroxyacetophenon
Formel:
C₈H₆F₂O₂ Schmelzpunkt: 105…109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042476 CAS-Nummer: 140675-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...