4-Amino-α,α,α-trifluor-o-toluylsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC1809-5GEA
14.7
EUR
APOSPC1809-5G
4-Amino-α,α,α-trifluor-o-toluylsäure 97%
4-Amino-α,α,α-trifluor-o-toluylsäure
Formel:
C₈H₆F₃NO₂ Molecular Weight: 205,14 g/mol Schmelzpunkt: 183…187 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407959 CAS-Nummer: 393-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...