2-(Brommethyl)-4-fluorphenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC1643-1GEA
315
EUR
APOSPC1643-1G
2-(Brommethyl)-4-fluorphenylboronsäure 95%
2-(Brommethyl)-4-fluorphenylboronsäure
Formel:
C₇H₇BBrFO₂ Schmelzpunkt: 169…171 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06659867 CAS-Nummer: 850568-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...