2-Amino-4'-fluorbenzophenon 95+%

Lieferant: Apollo Scientific

Achtung

APOSPC1569-100GEA 105 EUR
APOSPC1569-100G
2-Amino-4'-fluorbenzophenon 95+%
2-Amino-4'-fluorbenzophenon
Formel: C₁₃H₁₀FNO
Molecular Weight: 215,2230 g/mol
Schmelzpunkt: 124…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06658166
CAS-Nummer: 3800-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR