2-Chlor-5-ethoxyphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR9463-1GEA
123
EUR
APOSOR9463-1G
2-Chlor-5-ethoxyphenylboronsäure 97%
2-Chlor-5-ethoxyphenylboronsäure
Formel:
C₈H₁₀BClO₃ Schmelzpunkt: 98…100 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08235081 CAS-Nummer: 913835-30-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...