2-Amino-4,5-dimethylbenzoic acid

Lieferant: Apollo Scientific
Achtung

APOSOR941465-1GEA 80 EUR
APOSOR941465-1G
2-Amino-4,5-dimethylbenzoic acid
2-Amino-4,5-dimethylbenzoic acid
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Schmelzpunkt: 208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156998
CAS-Nummer: 15089-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR