3-Ethoxysalicylaldehyd 97%

Lieferant: Apollo Scientific

APOSOR937429-25GEA 69.8 EUR
APOSOR937429-25G
3-Ethoxysalicylaldehyd 97%
3-Ethoxysalicylaldehyd
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003323
CAS-Nummer: 492-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR