4-Hydroxydiphenylamin 95%
Lieferant: Apollo Scientific
APOSOR936320-25GEA
161
EUR
APOSOR936320-25G
4-Hydroxydiphenylamin 95%
4-Hydroxydiphenylamin
Formel:
C₁₂H₁₁NO Molecular Weight: 185,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020142 CAS-Nummer: 122-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...