4-Isobutylbenzoesäure 95%

Lieferant: Apollo Scientific

APOSOR921301-10GEA 401 EUR
APOSOR921301-10G APOSOR921301-5G APOSOR921301-1G
4-Isobutylbenzoesäure 95%
4-Isobutylbenzoesäure
Formel: C₁₁H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00191658
CAS-Nummer: 38861-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR