2-Cyclopentylidenessigsäure 98%
Lieferant: Apollo Scientific
APOSOR909192-1GEA
219
EUR
APOSOR909192-1G
APOSOR909192-5G
2-Cyclopentylidenessigsäure 98%
2-Cyclopentylidenessigsäure
Formel:
C₇H₁₀O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00034645 CAS-Nummer: 1903-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...