4-Heptylphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR7223-1GEA
253
EUR
APOSOR7223-1G
4-Heptylphenylboronsäure
4-Heptylphenylboronsäure
Formel:
C₁₃H₂₁BO₂ Schmelzpunkt: 81…83 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04039028 CAS-Nummer: 256383-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...