Phthalimidoacetaldehyddiethylacetal 98%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR6499-25GEA
87.3
EUR
APOSOR6499-25G
Phthalimidoacetaldehyddiethylacetal 98%
Phthalimidoacetaldehyddiethylacetal
Formel:
C₁₄H₁₇NO₄ Molecular Weight: 263,30 g/mol Schmelzpunkt: 72…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005901 CAS-Nummer: 78902-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...