4-Brom-N-(2-hydroxyethyl)benzolsulfonamid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
APOSOR5697-1GEA
33.5
EUR
APOSOR5697-1G
4-Brom-N-(2-hydroxyethyl)benzolsulfonamid 98%
4-Brom-N-(2-hydroxyethyl)benzolsulfonamid
Formel:
C₈H₁₀BrNO₃S Schmelzpunkt: 76…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03470703 CAS-Nummer: 59724-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...