5-Hydroxyisophthalsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Hydroxy-1,3-benzoldicarbonsäure

APOSOR4819-250GEA 161 EUR
APOSOR4819-250G APOSOR4819-1KG
5-Hydroxyisophthalsäure 98%
5-Hydroxyisophthalsäure
Formel: C₈H₆O₅
Molecular Weight: 182,13 g/mol
Schmelzpunkt: 298…302 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002515
CAS-Nummer: 618-83-7
EINECS: 210-565-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR