4-Bromo-2,2'-bipyridyl 95%

Lieferant: Apollo Scientific

APOSOR480861-1GEA 397 EUR
APOSOR480861-1G
4-Bromo-2,2'-bipyridyl 95%
4-Bromo-2,2'-bipyridyl
Formel: C₁₀H₇BrN₂
Molecular Weight: 235,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09910216
CAS-Nummer: 14162-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR