5-Methyl-2-pyridinboronsäure 95%

Lieferant: Apollo Scientific
Achtung

APOSOR4240-250MGEA 251 EUR
APOSOR4240-250MG APOSOR4240-1G
5-Methyl-2-pyridinboronsäure 95%
5-Methyl-2-pyridineboronic acid
Formel: C₆H₈BNO₂
Molecular Weight: 136,95
Schmelzpunkt: 86…90 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00674179
CAS-Nummer: 372963-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR