3-Chlor-4-(methylcarbamoyl)phenylboronsäure 95%

Lieferant: Apollo Scientific

Achtung

APOSOR4222-1GEA 132 EUR
APOSOR4222-1G
3-Chlor-4-(methylcarbamoyl)phenylboronsäure 95%
3-Chlor-4-(methylcarbamoyl)phenylboronsäure
Formel: C₈H₉BClNO₃
Molecular Weight: 213,43
Schmelzpunkt: 164…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363766
CAS-Nummer: 850589-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR