Ethyl-3-(3-boronobenzamido)propionat 96%
APOSOR4109-5GEA
0
EUR
APOSOR4109-5G
APOSOR4109-1G
Ethyl-3-(3-boronobenzamido)propionat 96%
Ethyl-3-(3-boronobenzamido)propionat
Formel:
C₁₂H₁₆BNO₅ Molecular Weight: 265,07 g/mol Schmelzpunkt: 128…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115701 CAS-Nummer: 850567-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...