1-Isopentyl-1H-pyrazol-4-boronsäurepinakolester 97%

Lieferant: Apollo Scientific

APOSOR4066-5GEA 549 EUR
APOSOR4066-5G APOSOR4066-1G
1-Isopentyl-1H-pyrazol-4-boronsäurepinakolester 97%
1-Isopentyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₄H₂₅BN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD05663867
CAS-Nummer: 777063-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR