2,3-Diphenylchinoxalin
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR40388-5GEA
42.3
EUR
APOSOR40388-5G
APOSOR40388-100G
APOSOR40388-25G
2,3-Diphenylchinoxalin
2,3-Diphenylquinoxaline
Formel:
C₂₀H₁₄N₂ Schmelzpunkt: 124…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014659 CAS-Nummer: 1684-14-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...