3-(2-Methoxyethylcarbamoyl)phenylboronsäure

Lieferant: Apollo Scientific

Achtung

APOSOR3983-5GEA 1050 EUR
APOSOR3983-5G APOSOR3983-1G
3-(2-Methoxyethylcarbamoyl)phenylboronsäure
3-(2-Methoxyethylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₄BNO₄
Schmelzpunkt: 130…132 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115710
CAS-Nummer: 850567-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR