4-Phenoxyphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

APOSOR3970-100GEA 261 EUR
APOSOR3970-100G APOSOR3970-25G
4-Phenoxyphenylboronsäure 98%
4-Phenoxyphenylboronsäure
Formel: C₁₂H₁₁BO₃
Molecular Weight: 214,03 g/mol
Schmelzpunkt: 141…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00093312
CAS-Nummer: 51067-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR