3-(Propylcarbamoyl)phenylboronsäure 95%
APOSOR3960-5GEA
405
EUR
APOSOR3960-5G
APOSOR3960-1G
3-(Propylcarbamoyl)phenylboronsäure 95%
3-(Propylcarbamoyl)phenylboronsäure
Formel:
C₁₀H₁₄BNO₃ Schmelzpunkt: 88…96 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04038919 CAS-Nummer: 850567-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...