4-(4-Bromphenylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

Achtung

APOSOR3787-1GEA 14.7 EUR
APOSOR3787-1G
4-(4-Bromphenylcarbamoyl)phenylboronsäure 98%
4-(4-Bromphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BBrNO₃
Molecular Weight: 319,95
Schmelzpunkt: 268…275 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027201
CAS-Nummer: 874287-99-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR