4-(Benzyl(ethyl)carbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR3772-1GEA
77.1
EUR
APOSOR3772-1G
4-(Benzyl(ethyl)carbamoyl)phenylboronsäure 98%
4-(Benzyl(ethyl)carbamoyl)phenylboronsäure
Formel:
C₁₆H₁₈BNO₃ Schmelzpunkt: 140…145 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09027215 CAS-Nummer: 913835-41-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...