4'-Methyl-4-biphenylboronic acid 97%

Lieferant: Apollo Scientific

APOSOR361428-1GEA 77.1 EUR
APOSOR361428-1G
4'-Methyl-4-biphenylboronic acid 97%
4'-Methyl-4-biphenylboronic acid
Formel: C₁₃H₁₃BO₂
Molecular Weight: 212,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038170
CAS-Nummer: 393870-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR