3-Chlor-5-hydroxyphenylboronsäure 95+%

Lieferant: Apollo Scientific

APOSOR360937-250MGEA 132 EUR
APOSOR360937-250MG
3-Chlor-5-hydroxyphenylboronsäure 95+%
3-Chlor-5-hydroxyphenylboronsäure
Formel: C₆H₆BClO₃
Molecular Weight: 172,3700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15143456
CAS-Nummer: 1214900-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR