4-Brom-2-nitrophenylboronsäure 95+%

Lieferant: Apollo Scientific

APOSOR360333-250MGEA 118 EUR
APOSOR360333-250MG
4-Brom-2-nitrophenylboronsäure 95+%
4-Brom-2-nitrophenylboronsäure
Formel: C₆H₅BBrNO₄
Molecular Weight: 245,8232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025992
CAS-Nummer: 860034-11-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR