2-Isopropoxy-5-pyridinboronsäure 95+%

Achtung

APOSOR360279-1GEA 0 EUR
APOSOR360279-1G APOSOR360279-5G APOSOR360279-10G
2-Isopropoxy-5-pyridinboronsäure 95+%
2-Isopropoxy-5-pyridinboronsäure
Formel: C₈H₁₂BNO₃
Molecular Weight: 181,00 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07368870
CAS-Nummer: 870521-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR