4-Formyl-3-methoxyphenylboronsäure 96%

Lieferant: Apollo Scientific

APOSOR360271-5GEA 908 EUR
APOSOR360271-5G APOSOR360271-1G
4-Formyl-3-methoxyphenylboronsäure 96%
4-Formyl-3-methoxyphenylboronsäure
Formel: C₈H₉BO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD10697427
CAS-Nummer: 815620-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR