2,4-Dimethoxy-5-pyrimidinboronsäurepinakolester 96%
Lieferant: Apollo Scientific
APOSOR360082-250MGEA
94.5
EUR
APOSOR360082-250MG
2,4-Dimethoxy-5-pyrimidinboronsäurepinakolester 96%
2,4-Dimethoxy-5-pyrimidinboronsäurepinakolester
Formel:
C₁₂H₁₉BN₂O₄ Molecular Weight: 266,1013 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06795654 CAS-Nummer: 936250-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...