3,4-(Methylenedioxy)phenylboronsäurepinakolester 95+%
Lieferant: Apollo Scientific
APOSOR360022-5GEA
109
EUR
APOSOR360022-5G
3,4-(Methylenedioxy)phenylboronsäurepinakolester 95+%
3,4-(Methylenedioxy)phenylboronsäurepinakolester
Formel:
C₁₃H₁₇BO₄ Molecular Weight: 248,0827 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663889 CAS-Nummer: 94838-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...