N-Benzyl-2-brombenzamid 98%

Lieferant: Apollo Scientific

APOSOR3595-1GEA 87.3 EUR
APOSOR3595-1G APOSOR3595-5G
N-Benzyl-2-brombenzamid 98%
N-Benzyl-2-brombenzamid
Formel: C₁₄H₁₂BrNO
Schmelzpunkt: 110…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00227668
CAS-Nummer: 82082-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR