2-Brom-N-isopropylbenzamid 98%

APOSOR3590-5GEA 0 EUR
APOSOR3590-5G APOSOR3590-1G
2-Brom-N-isopropylbenzamid 98%
2-Brom-N-isopropylbenzamid
Formel: C₁₀H₁₂BrNO
Molecular Weight: 242,1100 g/mol
Schmelzpunkt: 145…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00779656
CAS-Nummer: 64141-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR