2-(1-Adamantyl)-4-bromanisol 97%

Lieferant: Apollo Scientific

APOSOR3508-1GEA 151 EUR
APOSOR3508-1G
2-(1-Adamantyl)-4-bromanisol 97%
2-(1-Adamantyl)-4-bromanisol
Formel: C₁₇H₂₁BrO
Lagertemperatur: Raumtemperatur
MDL: MFCD03855308
CAS-Nummer: 104224-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR