4-Bromo-3-phenylpyrazole

Lieferant: Apollo Scientific

APOSOR350030-1GEA 66.9 EUR
APOSOR350030-1G
4-Bromo-3-phenylpyrazole
4-Bromo-3-phenylpyrazole
Formel: C₉H₇BrN₂
Molecular Weight: 223,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027561
CAS-Nummer: 13808-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR