1-Boc-3-indolboronsäurepinakolester

Lieferant: Apollo Scientific

APOSOR350029-1GEA 739 EUR
APOSOR350029-1G
1-Boc-3-indolboronsäurepinakolester
1-Boc-3-indolboronsäurepinakolester
Formel: C₁₉H₂₆BNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD12407262
CAS-Nummer: 942070-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR