4-(4-Chlorphenylcarbamoyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

APOSOR3497-1GEA 19 EUR
APOSOR3497-1G
4-(4-Chlorphenylcarbamoyl)phenylboronsäure 97%
4-(4-Chlorphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BClNO₃
Molecular Weight: 275,4953 g/mol
Schmelzpunkt: 248…250 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09027203
CAS-Nummer: 874288-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR