4-Isopropyl-3-thiosemicarbazid
Lieferant: Apollo Scientific
APOSOR346343-1GEA
94.5
EUR
APOSOR346343-1G
4-Isopropyl-3-thiosemicarbazid
4-Isopropyl-3-thiosemicarbazid
Formel:
C₄H₁₁N₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025144 CAS-Nummer: 13431-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...