N-Phenylglycinethylester
|
Achtung
|
APOSOR346330-25GEA
0
EUR
APOSOR346330-25G
APOSOR346330-100G
APOSOR346330-5G
N-Phenylglycinethylester
N-Phenylglycinethylester
Formel:
C₁₀H₁₃NO₂ Molecular Weight: 179,22 g/mol Schmelzpunkt: 56…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038315 CAS-Nummer: 2216-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...