5-Brom-o-toluonitril

Lieferant: Apollo Scientific

APOSOR345268-5GEA 61.1 EUR
APOSOR345268-5G
5-Brom-o-toluonitril
5-Brom-o-toluonitril
Formel: C₈H₆BrN
Molecular Weight: 196,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061920
CAS-Nummer: 156001-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR