3-Formyl-2-methoxyphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

APOSOR3445-1GEA 35 EUR
APOSOR3445-1G
3-Formyl-2-methoxyphenylboronsäure 98%
3-Formyl-2-methoxyphenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179,9700 g/mol
Schmelzpunkt: 122…125 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689486
CAS-Nummer: 480424-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR