3-Brom-(5-ethoxycarbonyl)phenylboronsäure 97%

APOSOR3440-1GEA 0 EUR
APOSOR3440-1G
3-Brom-(5-ethoxycarbonyl)phenylboronsäure 97%
3-Brom-(5-ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BBrO₄
Schmelzpunkt: 155…157 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689516
CAS-Nummer: 913835-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR