4-(2-Hydroxyethylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

APOSOR3427-5GEA 407 EUR
APOSOR3427-5G APOSOR3427-1G
4-(2-Hydroxyethylcarbamoyl)phenylboronsäure 98%
4-(2-Hydroxyethylcarbamoyl)phenylboronsäure
Formel: C₉H₁₂BNO₄
Molecular Weight: 209,01
Schmelzpunkt: 228…230 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659848
CAS-Nummer: 850593-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR