2-Chlor-(5-ethoxycarbonyl)phenylboronsäure 98%

Lieferant: Apollo Scientific

APOSOR3426-1GEA 183 EUR
APOSOR3426-1G
2-Chlor-(5-ethoxycarbonyl)phenylboronsäure 98%
2-Chlor-(5-ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BClO₄
Schmelzpunkt: 236…240 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689529
CAS-Nummer: 913835-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR