4'-Morpholinoacetophenon 95%
Lieferant: Apollo Scientific
APOSOR32584-5GEA
69.8
EUR
APOSOR32584-5G
4'-Morpholinoacetophenon 95%
4'-Morpholinoacetophenon
Formel:
C₁₂H₁₅NO₂ Molecular Weight: 205,2530 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006168 CAS-Nummer: 39910-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...