1-[(Ammoniooxy)methyl]-2,3-dichlorbenzolchlorid 95%

Lieferant: Apollo Scientific

APOSOR32397-1GEA 582 EUR
APOSOR32397-1G
1-[(Ammoniooxy)methyl]-2,3-dichlorbenzolchlorid 95%
1-[(Ammoniooxy)methyl]-2,3-dichlorbenzolchlorid
Formel: C₇H₇Cl₂NO·HCl
Molecular Weight: 228,504 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01114575
CAS-Nummer: 317821-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR